Anti-C17orf75 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1331AEA 407 CHF
ABGEAT1331A
Anti-C17orf75 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: C17orf75 (chromosome 17 open reading frame 75)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR