Anti-FRA10AC1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1325AEA 407 CHF
ABGEAT1325A
Anti-FRA10AC1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: FRA10AC1 (fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR