Anti-BUB1B Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1320AEA 407 CHF
ABGEAT1320A
Anti-BUB1B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: BUB1B (BUB1 mitotic checkpoint serine/threonine kinase B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR