Anti-BPNT1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1310AEA 407 CHF
ABGEAT1310A
Anti-BPNT1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: BPNT1 (3'(2'), 5'-bisphosphate nucleotidase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR