Anti-BCL2L12 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1283AEA 407 CHF
ABGEAT1283A
Anti-BCL2L12 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: BCL2L12 (BCL2-like 12 (proline rich))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR