Anti-BCAP31 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1277AEA 407 CHF
ABGEAT1277A
Anti-BCAP31 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: BCAP31 (B-cell receptor-associated protein 31)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR