Anti-B4GALNT1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1255AEA 407 CHF
ABGEAT1255A
Anti-B4GALNT1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: B4GALNT1 (beta-1,4-N-acetyl-galactosaminyl transferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR