Anti-B3GNT1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1254AEA 407 CHF
ABGEAT1254A
Anti-B3GNT1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: B3GNT1 (UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR