Anti-B3GALT2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1252AEA 407 CHF
ABGEAT1252A
Anti-B3GALT2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: B3GALT2 (UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR