Anti-ATP6V1G2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1243AEA 407 CHF
ABGEAT1243A
Anti-ATP6V1G2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ATP6V1G2 (ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR