Anti-ATP6V1C2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1242AEA 407 CHF
ABGEAT1242A
Anti-ATP6V1C2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ATP6V1C2 (ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR