Anti-ATP6V0D1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1240AEA 407 CHF
ABGEAT1240A
Anti-ATP6V0D1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ATP6V0D1 (ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR