Anti-ARPC2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1200AEA 407 CHF
ABGEAT1200A
Anti-ARPC2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ARPC2 (actin related protein 2/3 complex, subunit 2, 34kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR