Anti-ARID1B Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1189AEA 407 CHF
ABGEAT1189A
Anti-ARID1B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ARID1B (AT rich interactive domain 1B (SWI1-like))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR