Anti-ARHGAP6 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1180AEA 407 CHF
ABGEAT1180A
Anti-ARHGAP6 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ARHGAP6 (Rho GTPase activating protein 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR