Anti-AP3B1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1154AEA 407 CHF
ABGEAT1154A
Anti-AP3B1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AP3B1 (adaptor-related protein complex 3, beta 1 subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR