Anti-AMBP Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1129AEA 407 CHF
ABGEAT1129A
Anti-AMBP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AMBP (alpha-1-microglobulin/bikunin precursor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR