Anti-AKR7A2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1096AEA 407 CHF
ABGEAT1096A
Anti-AKR7A2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AKR7A2 (aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR