Anti-AKR1B10 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1092AEA 407 CHF
ABGEAT1092A
Anti-AKR1B10 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1B10 (aldo-keto reductase family 1, member B10 (aldose reductase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR