Anti-AKAP12 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1083AEA 407 CHF
ABGEAT1083A
Anti-AKAP12 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AKAP12 (A kinase (PRKA) anchor protein 12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR