Anti-AHSA1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1075AEA 407 CHF
ABGEAT1075A
Anti-AHSA1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AHSA1 (AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR