Anti-AEBP1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1062AEA 407 CHF
ABGEAT1062A
Anti-AEBP1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AEBP1 (AE binding protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR