Anti-ADH4 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1054AEA 407 CHF
ABGEAT1054A
Anti-ADH4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ADH4 (alcohol dehydrogenase 4 (class II), pi polypeptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR