Anti-ACTR2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1034AEA 407 CHF
ABGEAT1034A
Anti-ACTR2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ACTR2 (ARP2 actin-related protein 2 homolog (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR