Anti-ACTA2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1030AEA 407 CHF
ABGEAT1030A
Anti-ACTA2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR