Anti-ACAA2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1017AEA 407 CHF
ABGEAT1017A
Anti-ACAA2 Mouse Monoclonal Antibody
Type: Primary
Antigen: ACAA2 (acetyl-CoA acyltransferase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR