Anti-ABL2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1014AEA 407 CHF
ABGEAT1014A
Anti-ABL2 Mouse Monoclonal Antibody
Type: Primary
Antigen: ABL2 (c-abl oncogene 2, non-receptor tyrosine kinase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR