Anti-URGCP Rabbit Polyclonal Antibody [clone: 391CT6.4.3]

Supplier: Abgent
ABGEAP9984AEA 407 CHF
ABGEAP9984A
Anti-URGCP Rabbit Polyclonal Antibody [clone: 391CT6.4.3]
Antibodies
Type: Primary
Antigen: URGCP (upregulator of cell proliferation)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR