Anti-HIAT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9975BEA 405 CHF
ABGEAP9975B
Anti-HIAT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIAT1 (hippocampus abundant transcript 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR