Anti-ALG10 Rabbit Polyclonal Antibody [clone: 331CT17.4.4]

Lieferant: Abgent
ABGEAP9961AEA 405 CHF
ABGEAP9961A
Anti-ALG10 Rabbit Polyclonal Antibody [clone: 331CT17.4.4]
Antikörper
Type: Primary
Antigen: ALG10 (ALG10, alpha-1,2-glucosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR