Anti-DCT Rabbit Polyclonal Antibody [clone: 407CT20.1.2]

Supplier: Abgent
ABGEAP9956AEA 378 CHF
ABGEAP9956A
Anti-DCT Rabbit Polyclonal Antibody [clone: 407CT20.1.2]
Antibodies
Type: Primary
Antigen: DCT (dopachrome tautomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR