Anti-B3GAT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9926AEA 405 CHF
ABGEAP9926A
Anti-B3GAT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR