Anti-DDX6 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9900CEA 407 CHF
ABGEAP9900C
Anti-DDX6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX6 (DEAD (Asp-Glu-Ala-Asp) box helicase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR