Anti-AIG1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9898AEA 407 CHF
ABGEAP9898A
Anti-AIG1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AIG1 (androgen-induced 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR