Anti-DNAJC2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9885CEA 378 CHF
ABGEAP9885C
Anti-DNAJC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAJC2 (DnaJ (Hsp40) homolog, subfamily C, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR