Anti-PLB-T17 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9884AEA 378 CHF
ABGEAP9884A
Anti-PLB-T17 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLB-T17
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR