Anti-PXMP2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9872CEA 405 CHF
ABGEAP9872C
Anti-PXMP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PXMP2 (peroxisomal membrane protein 2, 22kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR