Anti-MAP4K3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9860AEA 448 CHF
ABGEAP9860A
Anti-MAP4K3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP4K3 (mitogen-activated protein kinase kinase kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR