Anti-HADHB Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9859BEA 407 CHF
ABGEAP9859B
Anti-HADHB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HADHB (hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR