Anti-GPR17 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9852CEA 378 CHF
ABGEAP9852C
Anti-GPR17 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPR17 (G protein-coupled receptor 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR