Anti-AGXT2 Rabbit Polyclonal Antibody [clone: 427CT2.1.1]
Lieferant: Abgent
ABGEAP9834BEA 405 CHF
ABGEAP9834B
Anti-AGXT2 Rabbit Polyclonal Antibody [clone: 427CT2.1.1]
Antikörper
Type: Primary
Antigen: AGXT2 (alanine--glyoxylate aminotransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human