Anti-PDK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9827AEA 405 CHF
ABGEAP9827A
Anti-PDK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PDK2 (pyruvate dehydrogenase kinase, isozyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR