Anti-NARFL Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9824CEA 405 CHF
ABGEAP9824C
Anti-NARFL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NARFL (nuclear prelamin A recognition factor-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR