Anti-PLA2G7 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9819CEA 405 CHF
ABGEAP9819C
Anti-PLA2G7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLA2G7 (phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR