Anti-SGMS2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9801BEA 448 CHF
ABGEAP9801B
Anti-SGMS2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SGMS2 (sphingomyelin synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR