Anti-HSD17B11 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9799AEA 407 CHF
ABGEAP9799A
Anti-HSD17B11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSD17B11 (hydroxysteroid (17-beta) dehydrogenase 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR