Anti-IDH3G Rabbit Polyclonal Antibody [clone: 434CT16.5.5]

Supplier: Abgent
ABGEAP9797BEA 407 CHF
ABGEAP9797B
Anti-IDH3G Rabbit Polyclonal Antibody [clone: 434CT16.5.5]
Antibodies
Type: Primary
Antigen: IDH3G (isocitrate dehydrogenase 3 (NAD+) gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR