Anti-LMBR1L Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9794BEA 405 CHF
ABGEAP9794B
Anti-LMBR1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMBR1L (limb development membrane protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR