Anti-GPI Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9786BEA 378 CHF
ABGEAP9786B
Anti-GPI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR