Anti-ATP6V1A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP9778BEA 407 CHF
ABGEAP9778B
Anti-ATP6V1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ATP6V1A (ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR